Please login to your online account to display your discounted pricing
Genlantis™ Dicer siRNA Generation Kits
Easily, economically generate functional siRNAs
$627.18 - $2,998.04
Specifications
For Use With (Application) Provides sufficient siRNAs in every single species by using ultra-active form of Human recombinant Dicer™ Enzyme to achieve effective gene silencing
View More Specs
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
T510001
genlantis
T510001
Recombinant Dicer™ Enzyme kit (50U), TurboScript™ T7 transcription kit (5 rxns), GeneSilencer™ siRNA transfection reagent kit (50 rxns), RNA purification column 1 and 2 50U Each for $2,173.84
T510002
genlantis
T510002
Recombinant Dicer™ Enzyme kit (0.5U/μL), Dicer™ Reaction Buffer, 50mM MgCl2 solution, 10mM ATP, Dicer™ stop solution, Nuclease-free Water 50U Each for $912.88
T510003
genlantis
T510003
T7 Enzyme mix, T7 Reaction Buffer, NTP mix, DNase I , 2X Gel Loading Buffer, Nuclease-free Water, LiCl precipitation solution, GFP control plasmid, 5' control primer, 3' control primer 20U Each for $627.18
T510008
genlantis
T510008
Recombinant Dicer™ Enzyme kit (0.5U/μL), Dicer™ Reaction Buffer, 50mM MgCl2 solution, 10mM ATP, Dicer™ stop solution, Nuclease-free Water 200U Each for $2,998.04
Description & Specifications
Specifications
For Use With (Application) Provides sufficient siRNAs in every single species by using ultra-active form of Human recombinant Dicer™ Enzyme to achieve effective gene silencing
  • Mixture of siRNAs has a better chance of success than a single siRNA design
  • More regions of the genes can be screened for silencing
  • Provided withthe proven GeneSilencer™ siRNA Transfection Reagent
  • Easy, cost-effective generation of a large number of small interfering RNAs (siRNAs) from full-length target genes
  • Purification columns available separately, see T51004 and T51005
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.