missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ Detachin™ Cell Detachment Reagent
Gentle, rapid detachment of a wide range of cells
Supplier: Genlantis™ T100110
Description
- Maximum cell viability
- Effective on a wide range of cells
- No mammalian or bacterial byproducts
- Stable at 4°C for two months
- No need to wash detached cells
- Suitable for detaching adherent cells from in vitro growth vesicles
- Provides quick and gentle detachment and dissociation of a wide variety of adherent cells, including primary cells, grown in all known tissue culture plasticware
Specifications
-20°C | |
Cell Culture Dissociation Reagent | |
10 x 100 mL |