Please login to your online account to display your discounted pricing

Genlantis™ Coomassie™ Stains


For rapid protein and DNA detection

Manufacturer: genlantis  D202025

 View more versions of this product

Catalog No. D202025

  • / Each
 

Description & Specifications

Specifications

Product Type Coomassie™ Brilliant blue G-250
Molecular Weight 854.04
Quantity 25g
Color Brilliant Blue
For Use With (Application) Protein and DNA detection