Please login to your online account to display your discounted pricing

Genlantis™ Coomassie™ Stains


For rapid protein and DNA detection

Manufacturer: genlantis  D203025

 View more versions of this product

Catalog No. D203025

  • / Each
 

Description & Specifications

Specifications

Product Type Coomassie™ Brilliant Blue R-250
Molecular Weight 854.04
Quantity 25g
Color Brilliant Blue
For Use With (Application) Protein and DNA detection
Grade Ultrapure
Type Coomassie™ Brilliant Blue R-250