Genlantis™ Bovine Smooth Muscle Cells
Economical alternative to primary human smooth muscle cells
Supplier: Genlantis™ PB35405
Description
- Each lot functionally characterized
- Same quality standard as the PrimaPure human cell products
- Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
- 16 population doublings guaranteed with Genlantis media and reagents
- Economical alternative suitable for investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Specifications
4°C | |
Bovine | |
>500000 Cells |
Bovine Aortic Smooth Muscle Cells (BAOSMC) | |
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells |