Please login to your online account to display your discounted pricing

Genlantis™ Bovine Smooth Muscle Cells


Economical alternative to primary human smooth muscle cells

Manufacturer: genlantis  PB35205

 View more versions of this product

Catalog No. PB35205

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Animal Bovine
Cell Type BPASMC
Quantity >500000 Cells
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells