Genlantis™ Bovine Smooth Muscle Cells
Economical alternative to primary human smooth muscle cells
Manufacturer:  Genlantis PB35005
 View more versions of this product
Catalog No. PB35005
Specifications Description & Specifications
Specifications
4°C
BCASMC
>500000 Cells
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Bovine