Genlantis™ Bovine Smooth Muscle Cells
Economical alternative to primary human smooth muscle cells
$1,412.20 - $1,416.33
Specifications
Animal Bovine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Quantity >500000 Cells
Storage 4°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Cell Type Price Quantity  
Catalog Number Mfr. No. Cell Type Price Quantity  
PB35205 Genlantis
PB35205
BPASMC Each for $1,412.20
PB35005 Genlantis
PB35005
BCASMC Each for $1,412.20
PB35405 Genlantis
PB35405
BAOSMC Each for $1,416.33
Specifications
Description
  • Each lot functionally characterized
  • Same quality standard as the PrimaPure human cell products
  • Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
  • 16 population doublings guaranteed with Genlantis media and reagents
  • Economical alternative suitable for investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Description & Specifications
Specifications
Bovine
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
>500000 Cells
4°C