Genlantis™ Bovine Smooth Muscle Cell Kits
Economical alternative to primary human smooth muscle cells
Supplier: Genlantis™ PB35205K
Specifications
4°C | |
Bovine | |
Cryopreserved cells, growth medium, subculture reagent kit |
Bovine Pulmonary Artery Smooth Muscle Cells (BPASMC) | |
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells | |
>500000 Cells |