Please login to your online account to display your discounted pricing

Genlantis™ Bovine Smooth Muscle Cell Kits


Economical alternative to primary human smooth muscle cells

Manufacturer: genlantis  PB35005K

 View more versions of this product

Catalog No. PB35005K

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Quantity >500000 Cells
Includes Cryopreserved cells, growth medium, subculture reagent kit
Animal Bovine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Cell Type BCASMC