Please login to your online account to display your discounted pricing

Genlantis™ Bovine Endothelial Cell Complete Kits


Economical alternative to human endothelial cells

Manufacturer: genlantis  PB30405

 View more versions of this product

Catalog No. PB30405

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Animal Bovine
Cell Type Aortic Cella (BAOEC)
For Use With (Application) To study endothelial function and endothelial metabolism, especially in co-culture of species matched bovine arterial smooth muscle cells
Quantity >500000 Cells