Please login to your online account to display your discounted pricing

Genlantis™ BoosterExpress™ Reagents and Kit


Up to 12 fold enhancement in gene expression levels post transfection

Manufacturer: genlantis  T20100B

 View more versions of this product

Catalog No. T20100B

  • / Each
 

Description & Specifications

Specifications

Quantity 3 x 1.5mL
Concentration 100X
For Use With (Application) Designed to significantly increase gene expression levels, efficient for both transient and stable transfections
Includes Reagents #1, #2 and #3 of 1 vial x 1.5mL