Genlantis™ BoosterExpress™ Reagents and Kit
Up to 12 fold enhancement in gene expression levels post transfection
$354.63 - $371.06
Specifications
For Use With (Application) Designed to significantly increase gene expression levels, efficient for both transient and stable transfections
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
T20102B Genlantis
T20102B
Reagent 2 Each for $371.06
T20101B Genlantis
T20101B
Reagent 1 Each for $371.06
T20103B Genlantis
T20103B
Reagent 3 Each for $371.06
T20100B Genlantis
T20100B
Reagents #1, #2 and #3 of 1 vial x 1.5mL Each for $354.63
Specifications
Description
  • Significantly enhanced gene expression levels in diverse cell lines
  • Compatibility with all major commercial transfection reagents including GenePORTER™ Transfection Reagents
  • Optimized conditions for popular cell lines
  • Efficient for both transient and stable transfections
  • Fast and easy protocol
  • Set of chemical cocktails designed to significantly increase gene expression levels
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Designed to significantly increase gene expression levels, efficient for both transient and stable transfections