missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genlantis™ Beta-Gal Assay Kits

Sensitive and accurate quantitation of β-Gal expression

$577.94 - $692.86

Specifications

Quantity 500 Assays
For Use With (Application) Sensitive and accurate quantitation of beta-gal expression
Storage Requirements -20°C
Sufficient For 500 Microassays in 96-well plate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
A10100K Genlantis
A10100K
5mL Lysis Buffer 5X, 55mL Standard Dilution Buffer, 55mL Substrate Buffer, 55mL Sto p Buffer, 5 x 1mL CPRG Substrate sto ck 10X, 100μL beta-gal Enzyme standard, 40U beta-Gal assay kit Each for $692.86
A10200K Genlantis
A10200K
55mL Lysis Buffer 5X, 55mL Standard Dilution Buffer, 55mL ONPG Substrate solution, 3mg/mL (o-nitrophenyl-beta-D-galacto pyranoside), 55mL Sto p Buffer, 100μL beta-gal Enzyme standard 40U FAQ B-galacto sidase kit Each for $577.94
Specifications

Description

  • Quantitative, sensitive, and convenient assays for determining βgalactosidase expression
  • Complete kits with ready-to-use reagents, including the E. coli βgalactosidase enzyme standard
  • Suitable for direct transfection assays in 96-well plates
  • Elimination of freeze-thaw cycles
  • The βGalactosidase Assay Kits quantitatively measure the expression level of βgalactosidase in cells transfected with vectors carrying the lacZ gene

Description & Specifications

Specifications

500 Assays
Sensitive and accurate quantitation of beta-gal expression
-20°C
500 Microassays in 96-well plate