Please login to your online account to display your discounted pricing
Genlantis™ BaculoPORTER™ Transfection Reagent
Novel lipid formulation optimized for insect cell transfection
Manufacturer: genlantis  T701035
 View more versions of this product
Catalog No. T701035
  • / Each
Description & Specifications
Specifications
Quantity 5 x 0.75mL
For Use With (Application) Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells