Genlantis™ BaculoPORTER™ Transfection Reagent
Novel lipid formulation optimized for insect cell transfection
Supplier:  Genlantis™ T701007
 View more versions of this product
Catalog No. T701007
Description
Description
A lipid formulation of a novel proprietary cationic lipid and dioleoyl phosphatidylethanolamine (DOPE) in a ratio targeted and optimized for the delivery of DNA into insect cells, especially Sf9 and Sf21.
  • Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells
  • Simple and straightforward protocol
  • Compatibility with diverse insect cell culture medias
  • Minimized cytotoxicity
Specifications
Specifications
Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells
1 x 0.75 mL