Genlantis™ BaculoPORTER™ Transfection Reagent
Novel lipid formulation optimized for insect cell transfection
$616.84 - $2,756.27
Specifications
For Use With (Application) Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T701035 Genlantis
T701035
5 x 0.75mL Each for $2,756.27
T701007 Genlantis
T701007
1 x 0.75mL Each for $616.84
Specifications
Description
A lipid formulation of a novel proprietary cationic lipid and dioleoyl phosphatidylethanolamine (DOPE) in a ratio targeted and optimized for the delivery of DNA into insect cells, especially Sf9 and Sf21.
  • Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells
  • Simple and straightforward protocol
  • Compatibility with diverse insect cell culture medias
  • Minimized cytotoxicity
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Highly efficient delivery of plasmids, bacmids and co-transfections consisting of a transfer vector and baculovirus DNA into Sf9 and Sf21 cells