Please login to your online account to display your discounted pricing

Genlantis™ 2EzTEV™ and 2EzHRV™ Dual-Tagged Proteases


Cleave fusion proteins at sites used in pGEX vectors

Manufacturer: genlantis  PEH0500

 View more versions of this product

Catalog No. PEH0500

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Recognizes the Peptide sequence Glu-Val-Leu-Phe-Gln-Gly-Pro and cleaves between the Gln-Gly residues
Concentration 2mg/mL
Storage Buffer 25mM Tris HCl, pH 8, 150mM NaCl, 14mM B-mecaptoethanol
Product Name 2EzHRV Dual-Tagged Protease
Molecular Weight 47
Storage Requirements -20°C
Quantity 500U
Source Human rhinovirus 3c
Includes Two purification tags (GST and 6xHis)