missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ 2EzTEV™ and 2EzHRV™ Dual-Tagged Proteases
Cleave fusion proteins at sites used in pGEX vectors
Supplier: Genlantis™ PET5000
Description
- Highly active, specific, and stable at the 4°C working temperature
- Do not require special buffer for activity and, therefore, can be used in the buffer that is most suitable for the target protein
- Contain two purification tags (GST and 6xHis), providing greater protein purification flexibility
2EzHRV Dual-Tagged Protease
- Genetically engineered cysteine protease from the human rhinovirus 3c
- Recognizes the peptide sequence Glu-Val-Leu-Phe-Gln-Gly-Pro and cleaves between the Gln-Gly residues
- MW: 47kDa
- Genetically engineered N1a protein (a cysteine protease) that contains the active catalytic fragment of the Tobacco Etch Virus (TEV)
- Recognizes the peptide sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between the Gln-Gly residues
- MW: 52kDa
Specifications
2 mg/mL | |
Recognizes the Peptide sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between the Gln-Gly residues | |
-20°C | |
2EzTEV Dual-Tagged Protease |
Two purification tags (GST and 6xHis) | |
25mM Tris HCl, pH 8, 150mM NaCl, 14mM B-mecaptoethanol | |
5000 U | |
52 |