missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genlantis™ 2EzTEV™ and 2EzHRV™ Dual-Tagged Proteases

Cleave fusion proteins at sites used in pGEX vectors

$638.73 - $1,277.49

Specifications

Product Name 2EzTEV Dual-Tagged Protease
Includes Two purification tags (GST and 6xHis)
Storage Buffer 25mM Tris HCl, pH 8, 150mM NaCl, 14mM B-mecaptoethanol
Storage Requirements -20°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Molecular Weight Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Molecular Weight Quantity Price Quantity  
PEH0500 Genlantis
PEH0500
Recognizes the Peptide sequence Glu-Val-Leu-Phe-Gln-Gly-Pro and cleaves between the Gln-Gly residues 47 500U Each for $638.73
PET5000 Genlantis
PET5000
Recognizes the Peptide sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between the Gln-Gly residues 52 5,000U Each for $1,277.49
Specifications

Description

  • Highly active, specific, and stable at the 4°C working temperature
  • Do not require special buffer for activity and, therefore, can be used in the buffer that is most suitable for the target protein
  • Contain two purification tags (GST and 6xHis), providing greater protein purification flexibility

2EzHRV Dual-Tagged Protease

  • Genetically engineered cysteine protease from the human rhinovirus 3c
  • Recognizes the peptide sequence Glu-Val-Leu-Phe-Gln-Gly-Pro and cleaves between the Gln-Gly residues
  • MW: 47kDa
2EzTEV Dual-Tagged Protease
  • Genetically engineered N1a protein (a cysteine protease) that contains the active catalytic fragment of the Tobacco Etch Virus (TEV)
  • Recognizes the peptide sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between the Gln-Gly residues
  • MW: 52kDa

Description & Specifications

Specifications

2EzTEV Dual-Tagged Protease
Two purification tags (GST and 6xHis)
25mM Tris HCl, pH 8, 150mM NaCl, 14mM B-mecaptoethanol
-20°C