Please login to your online account to display your discounted pricing

Genistein, Tocris


EGFR kinase inhibitor; Also estrogen and PPARγ ligand

Quantity 10mg

Manufacturer: r & d systems  RDS111010

 View more versions of this product

Catalog No. 111010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 270.24
Quantity 10mg
Chemical Name or Material Genistein
Assay Percent Range >98%
Molecular Formula C15H10O5
CAS 446-72-0
Synonym 5, 7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Genistein
Assay Percent Range >98%
Molecular Formula C15H10O5
Formula Weight 270.24
Synonym 5, 7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
Accessories

Desiccate at -20°C