missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genistein, 99%, synthetic, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 328270250

 View more versions of this product

Catalog No. AC328270250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Genistein
446-72-0
White to Beige
98.5% min. (HPLC)
C15H10O5
25 mg
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
TZBJGXHYKVUXJN-UHFFFAOYSA-N
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
5280961
270.2
Crystalline Powder
Synthetic
297.0°C to 298.0°C
Authentic
Glass bottle
MFCD00016952
15, 4426
Solubility in water: slightly soluble. Other solubilities: soluble in dmso and ethanol,soluble in usual organic solvents,soluble in diluted alkalies with yellow color
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
270.24
CHEBI:28088
99%

Chemical Identifiers

446-72-0
270.24
TZBJGXHYKVUXJN-UHFFFAOYSA-N
5280961
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O5
MFCD00016952
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
CHEBI:28088
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS Signal Word: Warning

EINECSNumber : 207-174-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only