Innovating Science™ Genetics of Blood Types Using Simulated Blood Kit
Learn how genetics determine blood type and possible inheritance patterns.
Manufacturer:  Innovating Science™ ADDD6502EMD
 View more versions of this product
Catalog No. S96791
Includes:
  • Four simulated blood samples
  • One bottle each anti-A and anti-B serum
  • 40 blood typing trays
Description
Description
Students use simulated blood to determine the blood type of four unknown samples and then use their results to assist in the resolution of a fictional paternity dispute.
  • Provides the most accurate simulation of the actual blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Hands-on activities
  • Kit contains materials for 10 groups
Uses only simulated blood. These activities do not use real blood.
Specifications
Specifications
49 CFR 173.4
White
14
7
4 simulated blood samples, mother sample x, child sample y, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 40 blood typing trays, Teacher's manual and student study guide copymasters
1
10 lab groups
13+ years
13
For fictional paternity test
Blood Typing Genetics Kit