Please login to your online account to display your discounted pricing
Genetic Traits Test Strips
A quick, easy, and completely safe means to analyze commonly tested genetic traits.
Manufacturer: fisher science education  AZ-60-5136
 View more versions of this product
Catalog No. S85289
  • / Pack of 100 Each
Description & Specifications
Specifications
For Use With (Application) To determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait
Product Type Thiourea Test Strips
Simple taste tests allow students to determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait.
Four types available:
  • PTC taste test strips
  • Sodium benzoate taste test strips
  • Thiourea taste test strips
  • Control taste test strips