Please login to your online account to display your discounted pricing
GeneJET DNA Purification Kits Promotion
Designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.
$103.18 - $103.18
For Use With (Application) Isolated genomic DNA is ideal for use in common molecular biology procedures, including: PCR and qPCR FastDigest and conventional restriction digestion Southern blotting
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
FERK0721PM
thermo scientific
PMK0721
50 Each for $103.18
Description & Specifications
Specifications
For Use With (Application) Isolated genomic DNA is ideal for use in common molecular biology procedures, including: PCR and qPCR FastDigest and conventional restriction digestion Southern blotting
Includes Proteinase K Solution, RNase A Solution, Digestion Solution, Lysis Solution, Wash Buffer I (concentrated), Wash Buffer II (concentrated), Elution Buffer, GeneJET Purification Columns (pre-assembled with collection tubes), Collection tubes, Detailed Protocol
Product Type Fermentas GeneJET DNA Purification Kit
  • Universal—can be used for both cell and tissue samples from a wide range of sources
  • Efficient—high yields of high molecular weight genomic DNA
  • Fast—20 minute procedure following cell lysis
  • Convenient—spin columns are capped and assembled with collection tubes