missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Geneclean™ Spin Glass Milk

For research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Manufacturer:  MP Biomedicals™ 111101401

 View more versions of this product

Catalog No. MP111101401



Description

Description

Usage Statement: Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications

Specifications

Store at 2-8°C
Kits and Assays
60mL
Cloudy heterogeneous, precipitate, off-white color
Documents