missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gellan Gum, Thermo Scientific Chemicals

An agar substitute and gelling agent

Supplier:  Thermo Scientific Chemicals J6342330

 View more versions of this product

Catalog No. AAJ6342330


Description

Description

Gellan Gum is an agar substitute and gelling agent, and useful in a wide variety of immobilization matricies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gellan Gum is an agar substitute and gelling agent. It is used in the food industry as an anti-settling agent, thickener or stabilizer. Also used in cosmetic industries due to its special gelling property. It is also used in pharmaceuticals and in tissue engineering. It is used as a substitute for agar in leptospiral media. Also useful in a wide variety of immobilization matricies.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

Gellan Gum
White
MFCD00131909
14,4383
Soluble in water.
71010-52-1
(C24H38O20)A
250 g
Phytagel
Powder

Chemical Identifiers

71010-52-1
(C24H38O20)A
MFCD00131909
Phytagel
Safety and Handling

Safety and Handling

EINECSNumber : 275-117-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only