Please login to your online account to display your discounted pricing

Alfa Aesar™ Geldanamycin, 1mg/mL in DMSO, sterile-filtered

Chemical Identifiers

Chemical Name or Material Geldanamycin
CAS 30562-34-6
Name Note 1mg/mL in DMSO, sterile-filtered
MDL Number MFCD00274570
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67124XF

 
alfa aesar™
J67124XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Geldanamycin
Physical Form Liquid
MDL Number MFCD00274570
CAS 30562-34-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Name Note 1mg/mL in DMSO, sterile-filtered
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1mL
RTECS Number LX8920000
Storage Note 1 -30° to -10°C
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

-30° to -10°C