Please login to your online account to display your discounted pricing

Gelatin Type A Type A; 225 Bloom Food Grade MP Biomedicals

$47.66 - $47.66

Chemical Identifiers

Chemical Name or Material Gelatin Type A
Name Note Type A; 225 Bloom
CAS 9000-70-8
Purity Grade Food Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN90177180

 
mp biomedicals
90177180
100g Each for $47.66
Description & Specifications

Specifications

Quantity 100g
Purity Grade Food Grade
Chemical Name or Material Gelatin Type A
CAS 9000-70-8
Name Note Type A; 225 Bloom