missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gelatin, type A, 175 Bloom

$57.46 - $204.85

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6154822 View Documents Alfa Aesar™
J6154822
100g Each for $57.46
 
AAJ6154836 View Documents Alfa Aesar™
J6154836
500g Each for $116.43
 
AAJ61548A1 View Documents Alfa Aesar™
J61548A1
1kg Each for $204.85
 
Description

Description

Applications
Gelatin is used to make lithographic and printing inks, plastic compounds, artificial silk, matches and pharmaceutical capsules. It finds application in light filters for mercury lamps, clarifying agent, pharmaceutical aid (suspending agent) and gelling component. It acts as an additive for foods and microencapsulating agent. Further, it is employed in electroplating, hectographic masters, paper and textile sizing, cosmetics, ointments and other medical and veterinary applications. It is useful in histochemistry, bacterial culture media and light microscope autoradiography. In addition to this, it is used to make the shells of paintballs and to hold silver halide crystals in an emulsion, in virtually all photographic films and photographic papers.

Solubility
Soluble in water, glycerol and acetic acid.

Notes
Of porcine originIncompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

9000-70-8
MFCD00081638

Specifications

White to Yellow
9000-70-8
14,4382
Gelatin, type A, 175 Bloom
100g
MFCD00081638
Powder
Safety and Handling

Safety and Handling

RTECSNumber : LX8580000

TSCA : TSCA