Please login to your online account to display your discounted pricing

Alfa Aesar™ Gelatin, 10% soln.

Chemical Identifiers

Chemical Name or Material Gelatin
CAS 9000-70-8
MDL Number MFCD00081638
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62699EQE

 
alfa aesar™
J62699EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gelatin
TSCA TSCA
Quantity 125mL
Physical Form Liquid
Concentration 10% soln.
MDL Number MFCD00081638
CAS 9000-70-8