Please login to your online account to display your discounted pricing
GE Osmonics™ Prefilter Syringe Filter
Manufacturer: maine manufacturing  MM1226946
 View more versions of this product
Catalog No. DGA04T30NB
  • / Pack of 500 Each
Description & Specifications
Specifications
No. per Pack 500/Pk.
Pore Size 0.45μm
Type Syringe Filter
Material Cellulose Acetate with Glass Fiber (CA/GF)
Applications Improve sample volume, filter viscous or high parliculate solutions