Please login to your online account to display your discounted pricing
GE Osmonics™ Prefilter Syringe Filter
Manufacturer: maine manufacturing  MM1226945
 View more versions of this product
Catalog No. DGA04T3050
  • / Pack of 50 Each
Description & Specifications
Specifications
Type Syringe Filter
No. per Pack 50/Pk.
Applications Improve sample volume, filter viscous or high parliculate solutions
Pore Size 0.45μm
Material Cellulose Acetate with Glass Fiber (CA/GF)