GE Osmonics™ Prefilter Syringe Filter
$237.72 - $1,880.47
Specifications
Type Syringe Filter
Material Cellulose Acetate with Glass Fiber (CA/GF)
Applications Improve sample volume, filter viscous or high parliculate solutions
Pore Size 0.45μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. No. per Pack Price Quantity  
Catalog Number Mfr. No. No. per Pack Price Quantity  
DGA04T3050 Maine Manufacturing
MM1226945
50/Pk. Pack of 50 Each for $237.72
DGA04T30NB Maine Manufacturing
MM1226946
500/Pk. Pack of 500 Each for $1,880.47
Specifications Description & Specifications
Specifications
Syringe Filter
Cellulose Acetate with Glass Fiber (CA/GF)
Improve sample volume, filter viscous or high parliculate solutions
0.45μm