Please login to your online account to display your discounted pricing

GE Osmonics™ Cellulose Acetate Syringe Filter


Manufacturer: maine manufacturing  MM1213641

 View more versions of this product

Catalog No. DDA02T3050

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Pore Size 0.22μm
Type Syringe Filter
Applications Used for maximum sample recovery
Material Cellulose Acetate (CA)
Diameter (Metric) 30mm