missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns
Designed for high performance, laboratory scale separations of proteins and other biomolecules based on size. GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns delivers quick and reproducible separations with high resolution.
$2,391.00 - $2,391.00
Specifications
Bed Volume | 24mL |
---|---|
Buffer | 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 1% SDS, Organic Solvents, 0.5M NaOH (for cleaning in-place) |
Diameter (English) Inner | 0.39 in. |
Diameter (Metric) Inner | 10mm |
For Use With (Application) | High performance gel filtration with a broad protein fractionation range of 5000 to 5,000,000 |
Includes: Valco fittings for AKTA™ design systems and are supplied with M6 connectors for FPLC System.
Catalog Number | Mfr. No. | Product Type | Flow Rate | Max. Pressure | Particle Size | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Flow Rate | Max. Pressure | Particle Size | Price | Quantity | ||
45-002-902 |
GE Healthcare
17517301 |
Superose 12 prepacked column | 0.5 to 1mL/min. | 435psi | 11μm |
Each for $2,391.00
|
|
Description
- Covers wide separation range of molecular weights from 1000 up to 5million MHighly cross-linked agarose matrixSmall average bead size of 13μm (Superose 6) and 11μm (Superose 12) are designed for laboratory-scale separations with good resolving capability
- Durable, high-performance glass columns for reliable and reproducible separations
- Available with two different media - Superose 6 or Superose 12
- Bed volume: 24mL
- Bed dimension: 10 × 300mm
- Pressure max.: 15 bar (217psi)
- pH stability working range: 3 to 12
- pH stability cleaning: 1 to 14
- Column I.D.: 10mm
- Bed support: filter
- Material (bed support): polypropylene
- Material (column hardware): borosilicate glass, PEEK, PP, EPDM and PFR
- Material (column tube): borosilicate glass
- Bed height with one adaptor: 279 to 309mm
- Bed height with two adaptors: 245 to 306mm
- Average partcle size: 13μm
- Gel filtration separations over a very broad molecular weight rang from 5000 to 5,000,000 (globular proteins)
- Flow rate: 0.3 to 0.5mL/min.
- Average partcle size: 11μm
- Gel filtration separations over a very broad molecular weight rang from 1000 to 300,000 (globular proteins)
- Flow rate of 0.5 to 1mL/min.
Media
Column
Superose 6 Media:
Superose 12 Media:
Specifications
24mL | |
0.39 in. | |
High performance gel filtration with a broad protein fractionation range of 5000 to 5,000,000 | |
300mm | |
Borosilicate Glass | |
4° to 30°C |
1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 1% SDS, Organic Solvents, 0.5M NaOH (for cleaning in-place) | |
10mm | |
11.81 in. | |
Valco fittings for AKTA™ design systems and are supplied with M6 connectors for FPLC System | |
3 to 12 | |
Cross-linked Agarose |