missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva (Formerly GE Healthcare Life Sciences) TiO2 Mag Sepharose™

Designed for enrichment of phosphopeptides from tryptic digested protein samples. Cytiva TiO2 Mag Sepharose magnetic beads are based on titanium dioxide chromatography and are designed to simplify capture and enrichment of phosphopeptides.

$64.61 - $691.45

Specifications

Bead Type TiO2 Mag Sepharose
Ligand TiO2
For Use With (Application) Used for selective capture and enrichment of Phosphopeptides, for biomarker research
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100μm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-002-158
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
28944010
500μL Each for $64.61
 
45-002-175
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
28951377
4 x 500μL Pack of 4 Each for $691.45
 
Description

Description

TiO2 Mag Sepharose has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. The magnetic bead format improves ease-of-use in sample preparation and has excellent properties for small-scale experiments. The high density beads allows rapid capture by magnetic devices with high visibility to ensure reliable collection of all bound target peptides.

  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel sample handling when used with MagRack 6 separation tool - process up to six samples in less than one hour

 

Specifications

Specifications

TiO2 Mag Sepharose
Used for selective capture and enrichment of Phosphopeptides, for biomarker research
37 to 100μm
TiO2
Paramagnetic spherical, highly cross linked agarose particles
4° to 30°C (use at room temperature)