missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Streptavidin Mag Sepharose™ Magnetic Bead

For simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules

$272.80 - $586.23

Specifications

Color White
Format Liquid
For Use With (Application) For simple and efficient enrichment of target Proteins by immunoprecipitation and purification of Biotinylated biomolecules
Storage Requirements 4° to 8°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-002-803 GE Healthcare
28985738
2 x 1mL Each for $272.80
45-002-804 GE Healthcare
28985799
5 x 1mL Each for $586.23
Specifications

Description

Streptavidin Mag Sepharose uses the strong interaction between biotin and streptavidin ligand, which is immobilized on magnetic beads.

  • High-capacity small-scale purification of proteins from cell lysates and biological fluids
  • Provides high purity and yield
  • Allows easy parallel screening of proteins with high repeatability
  • Simple enrichment of proteins from small or large sample volumes (low μl to high ml scale)
  • The beads are available in two pack sizes: 2 × 1mL 10% medium slurry and 5 × 1mL 10% medium slurry
  • A milliliter of 10% medium slurry is the same as 100 μl sedimented medium which is sufficient for 20 purification runs according to the recommended immunoprecipitation protocol
  • Binding capacity: >300μg biotinulated BSA/mL of medium slurry
  • Stability: stable

  • Immunoprecipitation
  • Purification
  • Biomarker reseach

Description & Specifications

Specifications

White
Liquid
For simple and efficient enrichment of target Proteins by immunoprecipitation and purification of Biotinylated biomolecules
4° to 8°C