Please login to your online account to display your discounted pricing

GE Healthcare StrepTactin™ Sepharose™ High Performance Medium

Mild purification conditions preserve antigenicity and function. GE Healthcare StrepTactin Sepharose High Performance media is used for robust, high resolution purification of Strep II-tagged proteins.

$931.00 - $931.00

Specifications

Product Type StrepTactin Sepharose HP media
Flow Rate <300 cm/hr.
For Use With (Application) Purification of Strep (II) -tagged recombinant Proteins to high purities in concentrated forms and small volumes by using with perstaltic pump or chromato graphy system
Ligand Type StrepTactin (∼5mg StrepTactin/mL medium)
Particle Size 34μm
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

45-001-585

 
ge healthcare
28935599
10mL Each for $931.00
Description & Specifications

Specifications

Product Type StrepTactin Sepharose HP media
Flow Rate <300 cm/hr.
For Use With (Application) Purification of Strep (II) -tagged recombinant Proteins to high purities in concentrated forms and small volumes by using with perstaltic pump or chromato graphy system
Ligand Type StrepTactin (∼5mg StrepTactin/mL medium)
Particle Size 34μm
pH Range 7
Storage Requirements 2° to 8°C
Support Type 6% Highly Cross-linked Agarose

  • Medium with high binding capacity yields high resolution, sharp concentrated peaks, and reproducible purifications
  • Physiological conditions and mild elution preserves the activity of the target protein
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Easily scaled up, and easily regenerated with 0.5M NaOH
  • Convenient use with a peristaltic pump or chromatography system

  • Matrix: rigid, highly cross-linked 6% agarose
  • Ave. particle size: 34μm
  • Dynamic binding capacity (protein dependent): approx. 6mg Strep(II)-tagged protein/mL medium
  • Compatibility: stable in all commonly used buffers, 0.5M NaOH (regeneration and cleaning), reducing agents, denaturants, and detergents
  • pH stability: >7