Please login to your online account to display your discounted pricing

GE Healthcare Standard Format HE33 Mini Submarine Unit and Accessories

Enhanced passive cooling system allows running gels at high voltages for speed and resolution

$490.00 - $593.00

Specifications

Amperage 500mA
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Dimensions (L x W x H) Exterior 5.2 x 9.5 x 2.8 in. (13 x 24 x 7cm)
Height (English) Exterior 2.8 in.
Height (Metric) Exterior 7cm
View More Specs
Includes:  Basic HE 33 Mini Submarine Unit:
Gel running tray, gel casting tray and bubble level

Complete HE 33 Mini Submarine Unit:
Basic unit, 1.5mm thick 8-well comb and comb back
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

45-001-852

 
ge healthcare
80605264
Gel running tray, gel casting tray and bubble level Mini submarine unit (Basic) Each for $490.00

45-001-851

 
ge healthcare
80605245
Basic unit, 1.5mm thick 8-well comb and comb back Mini submarine unit (Complete) Each for $593.00
Description & Specifications

Specifications

Amperage 500mA
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Dimensions (L x W x H) Exterior 5.2 x 9.5 x 2.8 in. (13 x 24 x 7cm)
Height (English) Exterior 2.8 in.
Height (Metric) Exterior 7cm
Length (English) Exterior 5.2 in.
Length (English) Gel 3.9 in.
Length (Metric) Exterior 13cm
Length (Metric) Gel 10cm
Volume (English) Buffer 8.4 oz.
Volume (Metric) Buffer 250mL
Width (English) Exterior 9.5 in.
Width (English) Gel 2.7 in.
Width (Metric) Exterior 24cm
Width (Metric) Gel 7cm
Sample Size Up to 32
Temperature (English) Operating 122°F
Temperature (Metric) Operating 50°C
Voltage 500V
Wattage 15w

  • Coolant-insulated buffer chamber runs 7 x 10cm agarose gels in <20 minutes
  • Convenient, leak-proof casting trays provide fast gel casting without tape
  • UV-transparent gel tray for convenient gel handling and viewing
  • Combs allow up to 32 samples per run
  • Molded base design to be filled with common coolant such as 50% ethylene glycol

For Basic unit, combs and comb back should be ordered separately