Please login to your online account to display your discounted pricing

Whatman™ SPARTAN™ HPLC Certified Syringe Filters


Purifies aqueous and organic solutions. Whatman™ SPARTAN™ HPLC Certified Syringe Filters are ready-to-use with a hydrophillic, low protein-binding membrane made of regenerated cellulose.

Manufacturer: whatman™  WHT10463050

 View more versions of this product

Catalog No. 09-302-153

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Color Code Light brown
Housing Material Polypropylene
Inlet Style Female luer lock
Quantity 100
Pore Size 0.45μm
Material Regenerated Cellulose (RC)
Outlet Style Male luer
Type Syringe Filter
Diameter (Metric) 30mm
Applications Filtration of organic and aqueous solutions in HPLC with reproducible results; Purification of aqueous and organic solutions; Filtration of protein solutions

SPARTAN syringe filters ensure reproducible results from the filtration of organic and aqueous solutions in HPLC. To ensure batch-to-batch consistency, the SPARTAN range of filters are tested and certified for the absence of UV-absorbing substances at wavelengths of 210 and 254nm with water, methanol and acetonitrile.

 

Features

  • Ready-to-use filter unit with a hydrophillic, low protein-binding membrane made of regenerated cellulose
  • Excellent chemical resistance against the standard aqueous and organic HPLC solvents
  • SPARTAN syringe filters are tested and certified for the absence of UV-absorbing substances at wavelengths of 210 and 254nm with water, methanol, and acetonitrile
  • 13mm diameter with Mini-Tip
  • Low dead volume (< 10μl)

Benefits

  • Versatile— Use for any application requiring a chemically resistant hydrophilic, low protein-binding membrane.
  • Documented batch-to-batch quality and consistency ensure reproducible results
  • Mini-Tip outlet is ideal for filtration into very small sample bottles
  • Filtration of organic and aqueous solutions in HPLC with reproducible results
  • Purification of aqueous and organic solutions
  • Filtration of protein solutions