Please login to your online account to display your discounted pricing
GE Healthcare SG Gradient Makers
Designed for producing linear gradients of aqueous solutions ranging in volume from 15 to 500mL
$645.81 - $915.67
Specifications
For Use With (Equipment) Vertical gel electrophoresis unit
Product Type Gradient Maker
Includes Ring stand support rod
Material Acrylic Plastic
View More Specs
Products
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity    
45-001-948
ge healthcare
80619761
0.5 oz. 15mL Each for $645.81
45-001-950
ge healthcare
80619799
1.69 oz. 50mL Each for $701.43
45-001-951
ge healthcare
80619818
16.9 oz. 500mL Each for $915.67
Description & Specifications
Specifications
For Use With (Equipment) Vertical gel electrophoresis unit
Product Type Gradient Maker
Includes Ring stand support rod
Material Acrylic Plastic
Choose from five sizes between 15mL and 500mL total volume.
  • Can be used to form convex or concave exponential gradients by the addition of a one-holed rubber stopper, a piece of rigid tubing, and a piece of flexible tubing
  • SG 50 and SG 100 units can be used to form convex and concave exponential gradients with the addition of an exponential gradient plunger accessory
  • The small units (SG 15, SG 30, SG 50, and SG 100) are machined from solid blocks of acrylic plastic
Acrylamide gel electrophoresis; Liquid chromatography elution; Sucrose, cesium, and preparative zonal centrifugation