Please login to your online account to display your discounted pricing

GE Healthcare Sephadex™ G15 Media

Gel filtration medium for desalting and buffer exchange for small biomolecules greater than 1500 molecular weight

$369.00 - $369.00

Products
Catalog Number Mfr. No. Price Quantity    

45-000-013

 
ge healthcare
17002001
Each for $369.00
Description & Specifications

  • For purposes of determining maximum flowrate and maximum pressure, beads behave as rigid spheres according to Darcy's Law
  • Working pH range 2 to 13
  • Particle size range for wet beads 60 to 180μm
  • Bed volume in distilled water 2.5 to 3.5mL/g
  • Exclusion limit: <1.5 x 103 for dextrans, <1.5 x 103 for globular proteins