Please login to your online account to display your discounted pricing

GE Healthcare Recombinant Taq DNA Polymerase

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

$510.88 - $510.88

Specifications

Format Liquid
For Use With (Application) PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Includes Taq DNA polymerase, 10x PCR buffer
pH 7.5
Product Name Recombinant Taq DNA Polymerase
View More Specs
Includes:  Taq DNA Polymerase, 10x PCR Buffer

 Disclaimers

This product has been reformulated such that it does not fall under the remaining claims of US patents 5,079,352, 5,789,224, 5,618,711, 6,127,155 and their foreign equivalents outside Canada. Therefore no license to practise under the aforementioned patents is conveyed by purchase of this product. No other patent rights (such as 5' Nuclease Process patent rights in US Patents Nos. 5,210,015 and 5,487,972) are conveyed expressly, by implication, or by estoppel.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

46-001-015

 
ge healthcare
28937345
1000U Each for $510.88
Description & Specifications

Specifications

Format Liquid
For Use With (Application) PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Includes Taq DNA polymerase, 10x PCR buffer
pH 7.5
Product Name Recombinant Taq DNA Polymerase
Source E. coli

Taq DNA Polymerase (cloned) is the recombinant form of the native enzyme from Thermus aquaticus expressed in E. coli.

  • Highly purified recombinant enzyme
  • Excellent batch-to-batch reproducibility
  • Each lot is tested to be free of nuclease contamination