Please login to your online account to display your discounted pricing
GE Healthcare PTFE-Coated Combs for Standard-Format Vertical Units
For use with Hoefer™ SE 600 Ruby™ vertical electrophoresis systems
$175.85 - $175.85
Specifications
Depth (English) 0.59 in.
Depth (Metric) 1.5cm
For Use With (Equipment) SE 600, SE 400 series vertical electrophoresis system
Product Type Comb
Material PTFE
View More Specs
Products
Catalog Number Mfr. No. Width (English) Well Thickness (English) Price Quantity    
45-001-909
ge healthcare
80-6163-41
4.4 in. 0.02 in. Each for $175.85
45-001-910
ge healthcare
80-6163-60
4.4 in. 0.03 in. Each for $175.85
45-001-912
ge healthcare
80-6164-17
4.7 in. 0.02 in. Each for $175.85
Description & Specifications
Specifications
Depth (English) 0.59 in.
Depth (Metric) 1.5cm
For Use With (Equipment) SE 600, SE 400 series vertical electrophoresis system
Product Type Comb
Material PTFE
No. of Wells 1 Preparative, 2 Reference
  • 25mm deep, adjustable to 10 or 15mm