missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Prostaglandin E2 Biotrak™ Assay

Enzyme immunoassay system suitable for plasma, urine, or cell culture supernatant samples

$745.00 - $745.00

Specifications

Assay Range 20 to 640pg/mL
For Use With (Application) ELISA
Total Assay Time 5 hr.
Sensitivity 16pg/mL
View More Specs

Includes: RPN222 supplied with sufficient quantities of reagents for 96 wells; RPN22210, for 960 wells. RPN22210 does not include lysis reagents. Both allow for construction of one standard curve and measurement of 37 unknowns in duplicate per plate.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
45-000-887 GE Healthcare
RPN222
96 wells Prostaglandin E2 Direct Biotrak assay Each for $745.00
45-001-312 GE Healthcare
RPN22210
10 x 96 wells Prostaglandin E2 Biotrak assay N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Specific for PGE2
  • Flexible method with choice of five assay protocols
  • Product available with or without lysis reagents
  • Lysis reagents facilitate extraction of PGEsubscript 2 from cell cultures and plasma samples; PGE is directly available for subsequent analysis without removal of extraction reagents
  • Precise and accurate measurement
  • Ready to use substrate and color-coded reagents
  • Range: 20-640pg/mL (1-32pg/well)
  • Room temperature protocol: 50-6400pg/mL
  • Sensitivity: 16pg/mL, 0.8pg/well
  • 5-hour protocol
  • Storage: Store at -15° to -30°C.
Description & Specifications

Specifications

20 to 640pg/mL
ELISA
5 hr.
16pg/mL