GE Healthcare Precision Column Holder
Adapts PC columns designed for SMART System and for use with other high performance chromatography systems including AKTAdesign systems
$984.00 - $984.00
Includes: Precision Column Holder, (Cat No. 45-000-253)
Column holder for 3-5cm columns, 1; Column holder for 30cm columns, 1; Screw caps, 4; End fittings, 4; Stop plugs, 4; Union M6 female 1/16 in. male 4
Column Holder, 10cm (Cat. No. 45-000-255)
Column holder for 10cm columns, 1; Screw caps, 2; End fittings, 2; Stop plugs, 2; Union M6 female 1/16 in. male, 2; Steering caps for steel column, 2
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
45-000-253 GE Healthcare
17145501
Each for $984.00
Specifications
Description
  • Low dead volume fittings maintain resolution
  • Includes all necessary tubing fittings and instructions