Please login to your online account to display your discounted pricing
GE Healthcare PlasmidSelect Xtra Chromatography Media
Used for generic purification of supercoiled plasmid DNA
$252.00 - $252.00
Products
Catalog Number Mfr. No. Price Quantity    
45-002-963
ge healthcare
28402401
Each for $252.00
Description & Specifications
  • Composed of 6% highly cross-linked agarose
  • Thiophilic aromatic adsorption media for consistent purification results when scaling up from research scale to cGMP production
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Available in convenient prepacked formats for research, process development and quantitative and qualitative analysis of supercoiled plasmid DNA