Please login to your online account to display your discounted pricing

GE Healthcare pI Markers

Run these markers in parallel with the experimental samples on isoelectric focusing gels. GE Healthcare Life Sciences™ pI Markers are lyophilized mixtures of stable, salt-free, highly purified proteins.

$374.00 - $394.74

Specifications

Color White
Quantity 10 Vials
Format Lyophilized
For Use With (Application) Isoelectric focusing
Storage Requirements 4°C
View More Specs
Products
Catalog Number Mfr. No. Kit Contents Product Type pH Range Price Quantity    

45-000-082

 
ge healthcare
17047201
Pepsinogen 2.80 pI (Native), Amylglucosidase 3.50 pI (Native), Methyl red (dye) 3.75 pI (Native), Glucose Oxidase 4.25 pI (Native), Trypsin inhibitor 4.55 pI (Native), bLactoglobulin A 5.20 pI (Native), Carbonic Anhydrase B (Bovine) 5.85 pI (Native) Low pI kit 2.8 to 6.55 Pack of 10 Each for $374.00

45-000-081

 
ge healthcare
17047101
Amylglucosidase - 3.50 pI (Native), Methyl red (dye) - 3.75 (pI Native), Trypsin inhibitor - 4.55 (pI Native), b-Lactoglobulin A - 5.20 (pI Native), Carbonic Anhydrase B (Bovine) - 5.85 (pI Native), Carbonic Anhydrase B (human) - 6.55 (pI Native) Broad pI kit 3 to 10 Pack of 10 Each for $394.74

45-000-083

 
ge healthcare
17047301
b-Lactoglobulin A - 5.20 pI (Native), Carbonic Anhydrase B (Bovine) - 5.85 pI (Native), Carbonic Anhydrase B (human) - 6.55 pI (Native), Myoglobin, Acidic - 6.85 pI (Native), Myoglobin, basic - 7.35 pI (Native), Lentil Lectin, Acidic - 8.15 pI (Native) High pI kit 5 to 10.5 Pack of 10 Each for $374.00
Description & Specifications

Specifications

Color White
Quantity 10 Vials
Format Lyophilized
For Use With (Application) Isoelectric focusing
Storage Requirements 4°C

The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.

  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c

Not for use in denaturing (urea) systems.