Please login to your online account to display your discounted pricing

GE Healthcare PhastGel™ Precast Gels

Select from homogeneous and gradient polyacrylamide gels. GE Healthcare Life Sciences™ PhastGel™ Precast Gels save time and improve reproducibility.

$185.00 - $426.00

Specifications

Color Colorless
Thickness (English) 0.017 in.
Width (Metric) Gel 5cm
For Use With (Application) Industrial applications such as analytical chemistry and research
Storage Requirements 4° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Formulation Height (Metric) Separation Range Concentration Price Quantity    

45-000-098

 
ge healthcare
17054501
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.8cm 5 to 8Mr 5 to 8% Pack of 10 Each for $204.00

45-000-097

 
ge healthcare
17054401
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.7cm 4 to 6.5Mr 6.5% Pack of 10 Each for $204.00

45-000-094

 
ge healthcare
17054001
Pack of 10 Each for $198.00

45-000-095

 
ge healthcare
17054201
Pack of 10 Each for $198.00

45-000-096

 
ge healthcare
17054301
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.6cm 3 to 9Mr 3 to 9% Pack of 10 Each for $204.00

45-000-119

 
ge healthcare
17062201
Pack of 10 Each for $185.00

45-000-121

 
ge healthcare
17062401
Pack of 10 Each for $185.00

45-000-127

 
ge healthcare
17067701
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.3cm Pack of 10 Each for $281.00

45-000-128

 
ge healthcare
17067801
Pack of 10 Each for $281.00

45-000-129

 
ge healthcare
17067901
High-density, 20% Polyacrylamide, 30% Ethylene Glycol 4.3cm 20 to 60Mr Pack of 10 Each for $426.00

45-000-120

 
ge healthcare
17062301
Pack of 10 Each for $185.00
Description & Specifications

Specifications

Color Colorless
Thickness (English) 0.017 in.
Width (Metric) Gel 5cm
For Use With (Application) Industrial applications such as analytical chemistry and research
Storage Requirements 4° to 8°C
Thickness (Metric) 0.04cm
Width (English) Gel 1.9 in.
Material Polyacrylamide

  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing, native, and isoelectric focusing gels
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling

  • PhastGel homogenous gels
    • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
    • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution
    PhastGel homogenous gels
    • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
  • PhastGel IEF gels
    • Homogeneous gels in pH ranges of 3-9, 5-8 and 4-6.5, for separations of proteins by isoelectric point